Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597521
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597521 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597521 Supplier Invitrogen™ Supplier No. PA597521
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COX5B Polyclonal Antibody for Western Blot, ICC/IF, ELISA

The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water. The mammalian COX apoenzyme is a dimer, with each monomer consisting of 13 subunits, some of which are mitochondrial and some of which are nuclear. Found in the inner mitochondrial membrane, COX5 is the heme A-containing chain of the oxidase family that converts one molecule of oxygen and four molecules of hydrogen to two molecules of water. Two isoforms of COX5 exist, COX5a and COX5b. When oxygen levels within the cell are high, transcription of COX5a (the aerobic isoform) is up-regulated as the rate of cellular respiration increases. Conversely, when oxygen levels are low, COX5b (the hypoxic isoform) transcription increases and functions to maximize the turnover rate of the COX apoenzyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX5B
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene COX5B
Gene Accession No. P10606
Gene Alias COX5B; COXVB; cytochrome c oxidase polypeptide Vb; cytochrome c oxidase polypeptide VB, mitochondrial; cytochrome c oxidase polypeptide VIa; cytochrome c oxidase precursor (EC 1.9.3.1); cytochrome c oxidase subunit 5B; cytochrome c oxidase subunit 5B, mitochondrial; cytochrome c oxidase subunit Vb; cytochrome c oxidase subunit VIA*
Gene Symbols COX5B
Host Species Rabbit
Immunogen Recombinant Human Cytochrome c oxidase subunit 5B, mitochondrial protein (32-129AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1329
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.