Learn More
Description
Coxsackievirus is a non-enveloped enterovirus that contains a linear, positive-sense single-stranded RNA.
- Propagated in the LLC-mk2 or Vero cell line
- Each lot is assayed for its tissue culture infective dose (TCID50) and sold with titers >105 U/mL
Specifications
Specifications
| Biosafety Level | 2 |
| Content And Storage | -65°C or below |
| For Use With (Application) | Nucleic acid/molecular testing, Limit of Detection (LOD) studies, Cross-reactivity studies, Other viral-based assays |
| Quantity | 1 mL |
Do not use in humans. For research use only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
