Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Coxsackievirus Type A8 Culture Fluid
SDP

Catalog No. 22157690 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-690 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains live and infectious organisms.
Catalog No. 22-157-690 Supplier ZeptoMetrix Supplier No. 0810143CF
Add to Cart
Add to Cart
Contains live and infectious organisms.

Infectious virus preparation designed to meet research and product development needs

Coxsackievirus is a non-enveloped enterovirus that contains a linear, positive-sense single-stranded RNA.

  • Consist of virus, cells and media taken directly from tissue culture flask
  • Propagated in RD cell line
  • Biosafety Level 2 organism
  • Available in 1.0mL aliquots
  • Each lot is assayed for its Tissue Culture Infective Dose (TCID50)
  • Titer: >105 U/mL

IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.

Nucleic Acid/Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays

Specifications

Biosafety Level 2
Certifications/Compliance ISO, 13485
Concentration >105U/mL
Content And Storage -65°C
Form Solution
Media Type Vial
Product Type Coxsackievirus Infectious Culture Fluid
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.