Learn More
Guardian™ CP-5, Chemical Resistant Smooth Finish 5 mil Butyl Gloves

Description
- Butyl material resists oxidation, ozone corrosion and abrasions
- Reusable and easy to clean
- Fitted hand style with smooth texture
- Protects against wide range of hazardous chemicals
- 5 mil thickness, 14 in. length
- Made in the USA
Specifications
Specifications
| Color | Black |
| Hand | Specific |
| Latex Free | Yes |
| Material | Butyl |
| Powdered | Powdered |
| Size | Small |
| Thickness (English) | 5 mil |
| Thickness (Metric) | 0.12 mm |
| Glove Cuff Style | Gauntlet |
| Grip Design | Smooth |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.