Learn More
Apexbio Technology LLC CP-91149(Synonyms: CP91149, CP 91149, Glycogen Phosphorylase Inhibitor CP-91149, GP inhibitor CP-91149), 10mg, CAS: 186392-40-5.
SDP
Supplier:  Apexbio Technology LLC A840310
Encompass_Preferred
CP-91149 (CAS 186392-40-5) is a selective inhibitor of glycogen phosphorylase (GP) an enzyme responsible for catalyzing glycogen breakdown into glucose-1-phosphate by cleavage of terminal -1 4-glycosidic bonds CP-91149 inhibits GP with an IC50 of 0 13 M In cell assays using primary human hepatocytes and isolated rat hepatocytes this inhibitor reduces glucagon-stimulated glycogenolysis (IC50 2 1 M and 10-100 M respectively) activates glycogen synthase and induces protein redistribution from soluble to particulate fractions mimicking insulin effects In vivo (ob/ob diabetic mice) CP-91149 treatment at 25 50 mg/kg rapidly decreases blood glucose without affecting normoglycemic animals underscoring its relevance in diabetes research
Catalog No. 50-000-60522
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.