Learn More
Selleck Chemical LLC CP-91149 S2717-100mg
SDP
Supplier:  Selleck Chemical LLC S2717100MG
Encompass_Preferred
CP-91149 is a selective glycogen phosphorylase (GP) inhibitor with IC50 of 0 13 M in the presence of glucose 5- to 10-fold less potent in the absence of glucose
Catalog No. 50-311-2512
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.