Learn More
Teledyne ISCO, Inc Cp Class Dual Piston Pump With Belt Drive And Shaped Cams; 0.0 - 100.0 Ml/Min.; 4,000 Psi
Supplier:  Teledyne ISCO, Inc CP100SRX
Encompass
CP Class Dual Piston Pump with Belt Drive and Shaped Cams; 0.0 - 100.0 mL/min.; 4,000 psi; Constant Pressure Algorithm; Stainless Steel Fluid Path; Automatic Piston Wash; Standard Seal (stainless steel energizer); Fitting package for 1/8"""" and 1/16"""" tubing
Catalog No. 50-211-9936
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.