Learn More
Specifications
Specifications
| Calibration Range | 0.78 to 50μg/mL |
| Conjugate | HRP |
| Cross Reactivity | Rat |
| Description | Cp (Rat) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of rat Cp in plasma and serum. |
| Detection Limit | 0.5μg/mL |
| Detection Method | Colorimetric |
| Includes | 96-well Antibody coated microwell, 25μg Rat Cp standard, 1 vial Biotinylated Cp, 30mL (10X) Mix diluent concentrate, 30mL Wash Buffer 20X, 90 Streptavidin-Peroxidase Conjugate (SP Conjugate), 8mL Chromogen Substrate, 12mL stop solution |
| Quantity | 1 Kit |
| Regulatory Status | For research use only (RUO) |
| Sample Type | Plasma, Serum |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.