Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CP110 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560139
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0139 Supplier Bethyl Laboratories, Inc Supplier No. A301343A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

CP110 is necessary for centrosome duplication at different stages of procentiole formation. It acts as a key negative regulator of ciliogenesis in collaboration with CEP97 by capping the mother centriole thereby preventing cilia formation. The diseases asssociated with CP110 inclued Spinocerebellar Ataxia 11 and Townes-Brocks Syndrome.

Specifications

Antigen CP110
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CCP110
Gene Accession No. O43303
Gene Alias centriolar coiled coil protein 110kDa, centriolar coiled-coil protein 110kDa, centrosomal protein CP110, centrosomal protein of 110 kDa, Cep110, CP110, KIAA0419
Gene Symbols CCP110
Host Species Rabbit
Immunogen Between 575 and 625
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9738
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.