Learn More
Selleck Chemical LLC CP21R7
SDP
Supplier:  Selleck Chemical LLC S79545MG
Encompass_Preferred
CP21R7 is a potent and selective GSK-3 inhibitor that can potently activate canonical Wnt signalling
Catalog No. 50-136-5473
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.