Learn More
TARGETMOL CHEMICALS INC CpCDPK1 TgCDPK1-IN-1 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T786051MG
Encompass_Preferred
Also available in 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. CpCDPK1/TgCDPK1-IN-1 is a potent dual inhibitor of CpCDPK1 and TgCDPK1, inhibits Abl and Src, and has antiparasitic activity for the study of Toxoplasma infections and cryptosporidiosis. Purity 98.71%
Catalog No. 50-287-9168
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.