Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CpG oligodeoxynucleotides (2216), Novus Biologicals™
SDP

Catalog No. NBP231136SS Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP231136SS 0.025 mg
NBP231136 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP231136SS Supplier Novus Biologicals Supplier No. NBP231136SS
Add to Cart
Add to Cart

Useful positive control model system for studying hTLR9 activation

Immunogen: G&asterisk;GGGGACGATCGTCG&asterisk;G&asterisk;G&asterisk;G&asterisk;G&asterisk;G Buffer: Sterile water Clone: 2216 Dilution: Ligand Activation, 5 to 20μg/mL CpG ODN Type A (2216)

Specifications

Concentration 1.0 mg/mL
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 0.025 mg
For Use With (Application) Human CpG ODNs are used for activation of human TLR9 at a range of 5 to 20μL. The TLR9/NF-kB/SEAPorter HEK 293 cell line is a useful positive control model system for studying hTLR9 activation. Typically use a 96-well plate format assay using this reporter cell line
Type CpG Oligodeoxynucleotides
Species Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.