Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CpG oligodeoxynucleotides Mouse
SDP

Catalog No. NBP231142 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP231142 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP231142 Supplier R&D Systems™ Supplier No. NBP231142
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.

Specifications

Concentration 1.0 mg/mL
Content And Storage Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Product Line Novus
Quantity 0.1 mg
For Use With (Application) Mouse CpG ODNs Are Used for Activation of Mouse TLR9 (at a Range of 5-20 μg/mL The TLR9/NF-kB/SEAPorter HEK 293 Cell Line is a Useful Positive Control Model System for Studying hTLR9 Activation. We Typically Use a 96-Well Plate Format Assay Using This Reporter Cell Line.)
Target CpG Oligodeoxynucleotides Mouse
Type CpG Oligodeoxynucleotides
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.