Learn More
Specifications
Specifications
| Concentration | 1.0 mg/mL |
| Content And Storage | Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.) |
| Product Line | Novus |
| Quantity | 0.025 mg |
| For Use With (Application) | Mouse CpG ODNs Are Used for Activation of Mouse TLR9 (at a Range of 5-20 μg/mL The TLR9/NF-kB/SEAPorter HEK 293 Cell Line is a Useful Positive Control Model System for Studying hTLR9 Activation. We Typically Use a 96-Well Plate Format Assay Using This Reporter Cell Line.) |
| Target | CpG Oligodeoxynucleotides Mouse |
| Type | CpG Oligodeoxynucleotides |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
