Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ CpGenom™ 5-hmC Quantitation Kit

Catalog No. 1710091 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-100-91 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-91 Supplier MilliporeSigma™ Supplier No. 1710091

missing translation for 'validMainframeMsg'

Includes

  • Plate Coating Buffer (15 mL)
  • 10X Assay Buffer (30 mL)
  • HRP Detection Reagent (15 mL)
  • 96-well Assay Plate and 8-well Strip Caps (1 plate/12 strips)
  • 5-hmC Antibody (1mg/mL)
  • 100x HRP Detection Antibody (1 mg/mL)
  • 5-hmC DNA Quantitation Standards - 5 vials for creation of standard curve (100 ng/μL each)

Requires

Incubator, thermal cyclers, Plate reader capable of 405 nm detection, Multichannel pipettor (recommended), Parafilm™, Pipette tips, dilution tubes, and other assorted consumables

Ideal for experiments where global and/or tissue-specific detection and quantitation of 5-hmC DNA is required with quantitation based on set of precisely calibrated standards

  • Compatible with genomic DNA from cultured cells or tissues
  • Includes set of 5-hmC of standards for precise quantitation
  • Streamlined, simple, automation-friendly ELISA-based protocol
  • Sensitive colorimetric detection with a limit of quantitation down to 0.03% of input DNA.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store components at the temperatures indicated on the labels
For Use With (Application) Determination of relative/absolute levels of 5-hydroxymethyl cytosine (5-hmC) in genomic DNA samples
Quantity 96-well Plate
Stability Kit components are stable for 6 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.