Learn More
MilliporeSigma™ CpGenome™ Direct Prep-96 Bisulfite Modification Kit
Catalog No. 1710454 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
192 Reactions
Catalog No. Quantity
17-104-54 192 Reactions
1 options
Catalog No. 17-104-54 Supplier MilliporeSigma™ Supplier No. 1710454
Add to Cart
Add to Cart
Requires
  • 96-100% Molecular Biology Grade Ethanol
  • Xylene (For FFPE Samples)
  • Thermal cycler, heat block, or water bath
  • Vortex or Eppendorf Thermomixer™
  • Microcentrifuge (capable of 12,000 x g)
  • Analytical scale or balance (For weighing tissue samples)
  • Untreated glass slides and slide washing chambers (For FFPE Samples)
Simple and reliable 96-well plate-based bisulfite conversion of DNA directly from a variety of starting materials, including cultured cells, blood, fresh tissue and fixed tissue samples.
Prep-96, Bisulfite, Modification, Kit
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) 96-well plate-based bisulfite conversion of DNA directly from a variety of starting materials, including cultured cells, blood, fresh tissue & fixed tissue samples
Quantity 192 Reactions