Learn More
Description
Includes
- 2X Extraction Buffer
- Modification Reagent
- Conversion Buffer
- Resuspension Buffer
- Equilibration Buffer
- Binding Buffer
- Wash Buffer I
- Wash Buffer II
- Elution Buffer
- Proteinase K
- Proteinase K Storage Buffer
- Desalting Columns
- Collection Tubes
Requires
MILLIPLEX MAP Cell Signaling Buffer and Detection Kit
- Bisulfite conversion directly from cells, tissues, blood and FFPE samples without DNA purification
- Sensitive bisulfite-conversion from as few as 10 cells or as low as 50pg of input DNA
- Fast and simple, streamlined protocol for one-step bisulfite conversion
- In-column desulfonation allows recovery of DNA without additional precipitation steps for more consistent results
- Suitable for downstream analysis by methlylation specific PCR, restriction digestion, sequencing, microarray hybridization, etc.
Specifications
Specifications
| For Use With (Application) | Bisulfite conversion directly from a variety of starting materials, including cultured cells, blood, fresh tissue & fixed tissue samples |
| Quantity | 50 Reactions |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.