Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam CPM, Thiol-Reactive Fluorescent Probe, >95% Purity, 50 mg

Catalog No. 502705344
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5344 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5344 Supplier Abcam Supplier No. AB14527550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Blue fluorescent thiol-reactive dye that becomes fluorescent upon reacting with thiols for non-separation quantification.

  • Molecular Weight: 402.44 g/mol
  • Purity: >95%
  • Properties: Widely used blue fluorescent thiol-reactive dye; non-fluorescent until reacted with thiols, allowing quantification without separation steps; acts as an energy acceptor from tryptophan and an energy donor to fluorescein.
  • Applications: Monitors thiol release and distinguishes proliferating cancer cells via nucleolar protein staining.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Thiols
Reactivity Thiols
Shipping Condition Ambient
Substrate Thiols
Label or Dye Fluorescent Probe
Product Type Protein Labeling Reagent
Detection Method Fluorescence
Buffer 7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin
Content And Storage Store at -20°C.
Detection Location In-Solution Detection
Formulation 100% 7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin
Quantity 50 mg
For Use With (Application) Quantifying thiols, nucleolar protein staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.