Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPN2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989442
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9442 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9442 Supplier Bioss Supplier No. BS9907R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CPN2, the 83 kDa subunit binds and stabilizes the catalytic subunit at 37 degrees Celsius and keeps it in circulation. Under some circumstances it may be an allosteric modifier of the catalytic subunit.

Specifications

Antigen CPN2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CPN2
Gene Accession No. P22792
Gene Alias 1300018K11Rik; ACBP; arginine carboxypeptidase; Carboxypeptidase N 83 kDa chain; Carboxypeptidase N large subunit; Carboxypeptidase N polypeptide 2; Carboxypeptidase N regulatory subunit; Carboxypeptidase N subunit 2; carboxypeptidase N, polypeptide 2; carboxypeptidase N, polypeptide 2 homolog; carboxypeptidase N, polypeptide 2, 83kD; CPN reg; CPN2; CPNreg; CPN-reg; RGD1305170
Gene Symbols CPN2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CPN2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1370
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.