Learn More
CPNE8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
The antibody has no cross reaction with CPNE5.
Specifications
Specifications
| Antigen | CPNE8 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.49 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CPNE8 |
| Gene Accession No. | Q86YQ8, Q9DC53 |
| Gene Alias | Copine 8, copine VIII, CPNE8 |
| Gene Symbols | Cpne8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.