Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPSF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555023
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555023 Supplier Invitrogen™ Supplier No. PA555023
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IELRKRVKLE GKELEEYLEK EKLKKEAAKK LEQSKEADID SSDESDIEED IDQPSAHKTK HDLMMKGEGS RKGSFFKQAK KSYPMFPAPE ERIKWDEYGE IIKPEDFLVP ELQATEEEKS KLESGLTNGD EPMDQDLSDV PTKCISTTES I Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.

CPSF2 gene ontology annotations related to this gene include mRNA cleavage; mRNA polyadenylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPSF2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CPSF2
Gene Accession No. O35218, Q9P2I0
Gene Alias 100kDa; 2610024B04Rik; AI662483; cleavage and polyadenylation specific factor 2; cleavage and polyadenylation specific factor 2, 100kD subunit; cleavage and polyadenylation specific factor 2, 100kDa; cleavage and polyadenylation specificity factor 100 kDa subunit; Cleavage and polyadenylation specificity factor subunit 2; Cpsf; CPSF 100 kDa subunit; CPSF 100kDa subunit; CPSF100; CPSF2; KIAA1367; Mcpsf; mKIAA1367
Gene Symbols CPSF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CPSF2. Recombinant protein control fragment (Product #RP-88985).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299256, 51786, 53981
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.