Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPSF3 Recombinant Rabbit Monoclonal Antibody (4S4P2)
GREENER_CHOICE

Catalog No. PIMA542768
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542768 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542768 Supplier Invitrogen™ Supplier No. MA542768
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: HeLa, 293T, SH-SY5Y, Mouse liver, Mouse kidney, Mouse uterus, Rat spleen. The target is usually found in the following locations: Nucleus. Immunogen sequence: LEVQSNPKIR KGAVQKVSKK LEMHVYSKRL EIMLQDIFGE DCVSVKDDSI LSVTVDGKTA NLNLETRTVE CEEGSEDDES LREMVELAAQ RLYEALTPVH.

Most eukaryotic mRNA precursors (pre-mRNAs) undergo extensive maturational processing, including cleavage and polyadenylation at the 3-prime end. CPSF3 encodes the 73-kD subunit of the cleavage and polyadenylation specificity factor (CPSF) and is the pre-mRNA 3-prime-end-processing endonuclease (Mandel et al., 2006).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPSF3
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4S4P2
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Cpsf3
Gene Accession No. Q9QXK7, Q9UKF6
Gene Alias cleavage and polyadenylation specific factor 3; cleavage and polyadenylation specific factor 3, 73kDa; cleavage and polyadenylation specificity factor 3; cleavage and polyadenylation specificity factor 73 kDa subunit; Cleavage and polyadenylation specificity factor subunit 3; CPSF; CPSF 73 kDa subunit; Cpsf3; Cpsf73; CPSF-73; mRNA 3'-end-processing endonuclease CPSF-73; YSH1
Gene Symbols Cpsf3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 585-684 of human CPSF3 (Q9UKF6).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298916, 51692, 54451
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.