Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPSF59 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560179
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0179 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0179 Supplier Bethyl Laboratories, Inc Supplier No. A301360A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Mouse

The function remains unknown.

Specifications

Antigen CPSF59
Applications Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene CPSF7
Gene Accession No. Q8N684
Gene Alias CFIm59, cleavage and polyadenylation specific factor 7, 59kDa, cleavage and polyadenylation specificity factor 59 kDa subunit, cleavage factor Im complex 59 kDa subunit, CPSF 59 kDa subunit, FLJ12529, pre mRNA cleavage factor I, 59 kDa subunit, pre-mRNA cleavage factor I, 59 kDa subunit, pre-mRNA cleavage factor Im 59 kDa subunit
Gene Symbols CPSF7
Host Species Rabbit
Immunogen Between 175 and 225
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79869
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.