Learn More
Cytiva (Formerly Pall Lab) GWV High Capacity Groundwater Sampling Capsules - 0.45 m (50/pkg)
Supplier:  Cytiva 12180
Encompass_Preferred
Superior Flow Rates and Higher Throughputs-75 Metals Analysis Certification is printed on each package.-Meets filtration requirements of the U.S. EPA. -Available in a variety of pore sizes to meet regional regulatory requirements.-Save time and money. Self-contained devices reduce the need for costly decontamination and multiple filter change steps associated with reusable filter holders.-Ensures rapid filtration. GWV provides five times the filtration area of conventional 142 mm filters. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3192
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.