Learn More
Cytiva AquaPrep™ Groundwater Sampling Capsules and Devices - 0.45 µm, Supor membrane (1/pkg)
Supplier:  Cytiva 12175
Encompass_Preferred
Optimized for Minimal Background in Dissolved Metals Analysis of Water samples-Easiest and most efficient way to meet filtration requirements of the U.S. EPA for 0.45 µm filtration.-Self-contained, disposable units eliminate the time and hazards associated with cleaning filter holders.-AquaPrep 600 capsule has over four times higher EFA than 142 mm disc filters, reducing the need for multiple filter changes during the sampling process and ensuring rapid sample filtration. -AquaPrep 600 capsules have Metals Ananlysis Certification on 48 metals printed on each package. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3189
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.