Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPT1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597615
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597615 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597615 Supplier Invitrogen™ Supplier No. PA597615
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CPT1A Polyclonal Antibody for Western Blot, IHC (P), ELISA

The mitochondrial oxidation of long-chain fatty acids is initiated by the sequential action of carnitine palmitoyltransferase I (which is located in the outer membrane and is detergent-labile) and carnitine palmitoyltransferase II (which is located in the inner membrane and is detergent-stable), together with a carnitine-acylcarnitine translocase. CPT I is the key enzyme in the carnitine-dependent transport across the mitochondrial inner membrane and its deficiency results in a decreased rate of fatty acid beta-oxidation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPT1A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Cpt1a
Gene Accession No. P50416, P97742
Gene Alias C730027G07; carnitine O-palmitoyltransferase; carnitine O-palmitoyltransferase 1, liver isoform; Carnitine O-palmitoyltransferase I, liver isoform; carnitine palmitoyl transferase I isoform; Carnitine palmitoyltransferase 1 alpha, liver isoform; carnitine palmitoyltransferase 1, liver; carnitine palmitoyltransferase 1A; carnitine palmitoyltransferase 1A (liver); carnitine palmitoyltransferase 1a, liver; carnitine palmitoyltransferase I, liver; CPT 1; CPT I; Cpt1; Cpt-1; CPT1 L; CPT1A; CPT1-L; CPTI; CPT-Ia; CPTI-L; L-CPT I; L-CPT1; mitochondrial carnitine palmitoyltransferase 1A
Gene Symbols Cpt1a
Host Species Rabbit
Immunogen Recombinant Human Carnitine O-palmitoyltransferase 1, liver isoform protein (301-600AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12894, 1374
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.