Learn More
Advanced ImmunoChemical, Inc. Recombinant canine parvovirus VP2
Supplier: Advanced ImmunoChemical, Inc. 8CPV20.5MG
Recombinant canine parvovirus variant 2b (CPV-2b) capsid protein VP2 consists of 584 amino acid residues. Theoretical molecular weight 64657 Da. Theoretical pI 5.36. Does not contain any affinity tags.