Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPXCR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541653
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541653 Supplier Invitrogen™ Supplier No. PA541653
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: WANQLAAVAA GARVAGTQAC ATETIDTSRV SLRAPQEFMT SHSEAGSRIV Sequence homology: Human: 100%.

The CPXCR1 gene is not extensively covered in the scientific literature. CXCL1, CXCR1, and CXCR2 are involved in chemotactic cytokine activities, which are crucial in inflammatory responses and conditions. CXCL1, for example, plays significant roles in angiogenesis and the recruitment of neutrophils under inflammatory conditions. Although CPXCR1 might be related due to its nomenclature and potential function in chemokine activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPXCR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CPXCR1
Gene Accession No. Q8N123
Gene Alias Cancer/testis antigen 77; CPX chromosomal region candidate gene 1 protein; CPX chromosomal region candidate gene 1 protein homolog; CPX chromosome region, candidate 1; Cpxcr1; CT77; Gm1143; hypothetical LOC100360571; LOC100360571
Gene Symbols CPXCR1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CPXCR1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53336
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.