Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRACC Monoclonal Antibody (5G5A4)
GREENER_CHOICE

Catalog No. FERMA548769
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548769 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548769 Supplier Invitrogen™ Supplier No. MA548769
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CRACC Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

SLAMF7 contains one Ig-like C2-type (immunoglobulin-like) domain. Isoform 1 mediates NK cell activation through a SAP-independent extracellular signal-regulated ERK-mediated pathway. It may play a role in lymphocyte adhesion. Isoform 3 does not mediate any activation. SAP can bind the cytoplasmic tail of isoform 1 when phosphorylated in the presence of Fyn (in vitro). SLAMF7 is expressed in spleen, lymph node, peripheral blood leukocytes, bone marrow, small intestine, stomach, appendix, lung and trachea. Expression was detected in NK cells, activated B-cells, NK-cell line but not in promyelocytic, B-, or T-cell lines. The isoform 3 is expressed at much lower level than isoform 1. There are three named isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRACC
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone 5G5A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene SLAMF7
Gene Accession No. Q9NQ25
Gene Alias 19A; 19A24; 19A24 protein; 4930560D03Rik; CD2 subset 1; CD2-like receptor activating cytotoxic cells; CD2-like receptor-activating cytotoxic cells; CD319; CRACC; CS1; Leukocyte cell-surface antigen; membrane protein FOAP-12; Novel Ly9; novel LY9 (lymphocyte antigen 9) like protein; protein 19A; RGD1560190; SLAM family member 7; SLAMF7; UNQ576/PRO1138
Gene Symbols SLAMF7
Host Species Mouse
Immunogen Purified recombinant fragment of human CD319 (AA: extra 23-226) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57823
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.