Learn More
CPC Scientific H-Gly-Leu-Leu-Arg-Lys-Gly-Gly-Glu-Lys-Ile-Gly-Glu-Lys-Leu-Lys-Lys-Ile-Gly-Gln-Lys-Ile-Lys-Asn-Phe-Phe-Gln-Lys-Leu-Val-Pro-Gln-Pro-Glu-Gln-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific CRAM001B
Encompass
CRAMP is the first antibiotic peptide found in cells of myeloid lineage in the mouse, a potent antibiotic against Gram-negative bacteria.ONE-LETTER SEQUENCE: GLLRKGGEKIGEKLKKIGQKIKNFFQKLVPQPEQMOLECULAR FORMULA: C178H302N50O46MOLECULAR WEIGHT:3878.67STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [376364-36-2], RESEARCH AREA: AntimicrobialREFERENCES: L.Saiman et al., Antimicrob. Agents Chemother., 45, 2838 (2001); P.Bergman et al., Infect. Immun., 74, 6982 (2006)
Catalog No. 50-210-9158