Learn More
Cayman Chemical CRAMP140-173mousetrIf 10mg
SDP
Catalog No. 502917385
Encompass_Preferred
An antimicrobial peptide; active against K. pneumoniae in vitro at 1 UG/ml; decreases neutrophil infiltration of colonic epithelial and luminal surfaces in a porcine model of colitis at 10 mg/kg; ileal administration decreases disease severity and cecal bacterial burden and increases survival in a mouse model of C. difficile infection at 10 mg/kg; intra-articular administration increases disease severity and induces synovitis in a mouse model of surgery-induced osteoarthritis
Catalog No. 50-291-7385 Supplier Cayman Chemical Supplier No. 3750210 MG
An antimicrobial peptide; active against K. pneumoniae in vitro at 1 UG/ml; decreases neutrophil infiltration of colonic epithelial and luminal surfaces in a porcine model of colitis at 10 mg/kg; ileal administration decreases disease severity and cecal bacterial burden and increases survival in a mouse model of C. difficile infection at 10 mg/kg; intra-articular administration increases disease severity and induces synovitis in a mouse model of surgery-induced osteoarthritis