Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Cranad-2 Fluorescent Probe, Amyloid-beta, Solid, 5 mg

Catalog No. 502706425
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6425 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6425 Supplier Abcam Supplier No. AB1417755MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Near-infrared fluorescent probe for in vitro and in vivo detection of amyloid-beta aggregates.

  • Molecular Weight: 410.3 Da
  • Purity: >98%
  • Form: Solid
  • Properties: Near-infrared (NIR) fluorescent probe for Amyloid-β aggregates. Displays a 70-fold increase in fluorescence intensity upon interaction with Amyloid-β aggregates, accompanied by a 90 nm blue shift (805 to 715 nm).
  • Binding Affinity: High affinity for Amyloid-β aggregates (Kd = 38 nM).
  • Applications: Detects Amyloid-β in vitro and in vivo. Blood-brain barrier permeable.
  • Storage: Store at -20°C.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Amyloid-β
Substrate Properties Chemical Substrate
Substrate Amyloid-beta
Label or Dye Fluorescent Probe
Shipping Condition Blue Ice
Content And Storage Store at -20°C.
Form Solid
Detection Method Fluorescence
Cross Reactivity Amyloid-β aggregates
Quantity 5 mg
For Use With (Application) Amyloid-beta imaging
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.