Learn More
Abcam Cranad-2, Near-infrared Amyloid-beta Fluorescent Probe, Solid, 25 mg
Description
- Molecular Weight: 410.3 Da
- Purity: >98%
- Physical Form: Solid
- Properties: Near-infrared (NIR) fluorescent probe for Amyloid-β aggregates; displays a 70-fold increase in fluorescence intensity and a 90 nm blue shift from 805 to 715 nm upon binding.
- Affinity: High affinity for Amyloid-β aggregates (Kd = 38 nM).
- Applications: Detects Amyloid-β in vitro and in vivo; blood-brain barrier permeable.
- Storage: Store at -20°C.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Amyloid-β (Aggregates) |
| Substrate Properties | Chemical Substrate |
| Substrate | Amyloid-beta aggregates |
| Shipping Condition | Blue Ice |
| Label or Dye | Fluorescent Probe |
| Form | Solid |
| Content And Storage | Store at -20°C. |
| Detection Method | Fluorescence |
| Cross Reactivity | Amyloid-beta |
| Quantity | 25 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.