Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRASP-2 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990902
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0902 100 μg
50-199-0903 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0902 Supplier Rockland Immunochemicals Supplier No. 200401C19
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for Borrelia burgdorferi CRASP-2 protein. A BLAST analysis was used to suggest reactivity with CRASP-2 from B. burgdorferi sources based on 100% homology with the immunizing sequence. Partial cross-reactivity is expected against B. garinii, B. spielmanii, and valaisiana sources based on 91-89% homology. Cross-reactivity with CRASP-2 from other sources has not been determined.

CRASP-2 (Complement Regulator-Acquiring Surface Protein 2) of Borrelia burgdorferi binds FHL-1 and factor H binding protein in a distinct way. It may be predominantly expressed by serum-resistant Borrelia strains. Borrelia burgdorferi sensu lato has the ability to evade immune systems to persist in a variety of vertebrate hosts. This activity is dependent on a number of factors. Some Borrelia species bind host-derived fluid-phase immune regulators FHL-1 and factor H to their surface via complement regulator-acquiring surface proteins (CRASPs). Factor H and FHL-1 serve as cofactors for factor I, a serine protease that cleaves complement component 3b (C3b) directly on the cell surface and thereby confers resistance of spirochetes to complement-mediated lysis. It is possible that because of discontinuous binding regions in the factor H/FHL-1, long distance interaction may be involved in binding of both immune regulators. Putative coiled-coil structural elements may be important in the interaction of B. burgdorferi CRASP-1 with factor H.

Specifications

Antigen CRASP-2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Immunogen MBP-fusion protein corresponding to Borrelia burgdorferi CRASP-2 protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.