Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRBN Mouse anti-Human, Unconjugated, Clone: 4D6 (-3H10-D6), MilliporeSigma™

Catalog No. MABC15931MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC15931MI 100 μg
MABC15932MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC15931MI Supplier MilliporeSigma Supplier No. MABC1593100UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

CRBN Monoclonal antibody specifically detects CRBN in Human samples. It is validated for Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRBN
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4D6 (-3H10-D6)
Conjugate Unconjugated
Gene Accession No. NP_001166953.1
Gene Alias Protein Cereblon;CRBN
Gene Symbols CRBN
Host Species Mouse
Immunogen His-tagged recombinant protein fragment corresponding to the N-terminal region of human Cereblon.
Molecular Weight of Antigen ∼50 kDa observed; 50.55 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity Clone 4D6 (-3H10-D6) is a mouse monoclonal antibody that detects human Cereblon. It targets an epitope within the N-terminal domain.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.