Learn More
CRBN Mouse anti-Human, Unconjugated, Clone: 4D6 (-3H10-D6), MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | CRBN |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 4D6 (-3H10-D6) |
| Conjugate | Unconjugated |
| Gene Accession No. | NP_001166953.1 |
| Gene Alias | Protein Cereblon;CRBN |
| Gene Symbols | CRBN |
| Host Species | Mouse |
| Immunogen | His-tagged recombinant protein fragment corresponding to the N-terminal region of human Cereblon. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.