Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRBN Recombinant Rabbit Monoclonal Antibody (PSH0-69)
GREENER_CHOICE

Catalog No. FERMA549359 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA549359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA549359 Supplier Invitrogen™ Supplier No. MA549359
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Sequence Similarities: 95% Mouse/Rat. Tissue Specificity: Widely expressed. Highly expressed in brain. Positive Control: HEK-293 cell lysate, A375 cell lysate, A549 cell lysate, HeLa cell lysate, SH-SY5Y cell lysate, NIH/3T3 cell lysate, MCF7 cell lysate, mouse brain tissue lysate, rat testis tissue lysate, human testis tissue, human brain tissue. Subcellular Location: Cytoplasm, Nucleus, Membrane. Predicted band size: 51 kDa.

CRBN, a member of the Lon protease protein family, plays a role in brain development. It is a component of the DCX (DDB1-CUL4-X-box) E3 protein ligase complex, a complex that mediates the ubiquitination and subsequent proteasomal degradation of target proteins and is required for limb outgrowth and expression of the fibroblast growth factor FGF8. CRBN is thought to regulate the assembly and neuronal surface expression of large-conductance calcium-activated potassium channels in brain regions involved in memory and learning via its interaction with KCNT1. It is widely expressed and highly expressed in brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRBN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone PSH0-69
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CRBN
Gene Accession No. Q56AP7, Q8C7D2, Q96SW2
Gene Alias 2610203G15Rik; 2900045O07Rik; AD-006; AF229032; AW108261; cereblon; CRBN; DKFZp781K0715; MGC27358; MRT2; MRT2A; novel lethal; piL; protein cereblon; Protein PiL; protein x 0001
Gene Symbols CRBN
Host Species Rabbit
Immunogen Recombinant protein within human crbn aa 1-442 / 442.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297498, 51185, 58799
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.