Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRCP Recombinant Rabbit Monoclonal Antibody (JE65-42)
GREENER_CHOICE

Catalog No. PIMA544746
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544746 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544746 Supplier Invitrogen™ Supplier No. MA544746
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CRCP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes a membrane protein that functions as part of a receptor complex for a small neuropeptide that increases intracellular cAMP levels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRCP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE65-42
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CRCP
Gene Accession No. O35427, O75575
Gene Alias AL022669; calcitonin gene-related peptide-receptor component protein; CGRP receptor component; CGRPRCP; CGRP-RCP; CGRP-receptor component protein; Crcp; DNA-directed RNA polymerase III subunit RPC9; RCP; RCP9; RNA polymerase III subunit C9
Gene Symbols CRCP
Host Species Rabbit
Immunogen Synthetic peptide within human CRCP aa 10-60/148.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12909, 27297
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.