Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cre Recombinase Mouse anti-Virus, Clone: 1D8H3, GenScript™

Catalog No. 50273029
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-273-029 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-273-029 Supplier Genscript Corporation Supplier No. A0088840
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/mL. Cre-recombinase is a 38 kDa DNA recombinase derived from the P1 bacteriophage. It is highly specific to a 34 bp DNA sequence (loxP) found in P1 DNA. It catalyzes site-specific recombination between two 34-base-pair LOXP sites. Its role is to maintain the phage genome as a monomeric unit-copy plasmid in the lysogenic state. It is homotetramer when bound to DNA and belongs to the phage integrase family.GenScript Mouse Anti-Cre recombinase Monoclonal Antibody is an important tool for the identification of recombinant Cre recombinase from E.coli expression systems.

Cre Recombinase Monoclonal antibody specifically detects Cre Recombinase in Virus samples. It is validated for ELISA, Western Blot

Specifications

Antigen Cre Recombinase
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D8H3
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene Alias Cre
Host Species Mouse
Immunogen Purified recombinant Cre recombinase protein
Purification Method Protein A
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.