Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatine Deiminase from Microbial , MP Biomedicals, LLC

Catalog No. MP215349025 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215349025 25U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215349025 Supplier MP Biomedicals, Inc Supplier No. 0215349025
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Creatinine deiminase has been used in a study to assess the application of a creatinine-sensitive biosensor for hemodialysis control.

  • Creatinine deiminase has also been used in a study to investigate the bioelectronic tongue for the simultaneous determination of urea, creatinine and alkaline ions in clinical samples
  • One unit causes the formation of oneμmole of ammonia (the oxidation of oneμmole of NADPH) per minute at pH 7.5 at 37°C
TRUSTED_SUSTAINABILITY

Specifications

Color White
pH 8.5 to 9.5 (Optimum)(literature) pH Stability: 7.0 to 11.0 (30°C 20hr)
Packaging Glass
Quantity 25U
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.