Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Creatine Kinase-MB Kit, Calibrator Grade Recombinant Protein

Catalog No. PIAKC0315
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIAKC0315 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIAKC0315 Supplier Invitrogen™ Supplier No. AKC0315
Add to Cart
Add to Cart

Recombinant Protein

Kit includes 100 μg of lyophilized recombinant Human CK-MB protein and 1 bottle of diluting buffer (1 mL). Storage: Diluent should be stored at 4°C. Lyophilized protein should be stored at 4°C short-term, and long-term at -20°C. Reconstitution: reconstitute 100 μg of Recombinant Human CK-MB, Calibrator Grade with 1 mL of CK-MB Diluent. Mix gently, and allow to stand for at least 15 minutes prior to use. After using, replace stopper and store at 2-8oC to achieve at least 3 weeks of reconstituted stability.

The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P06732, P12277
For Use With (Application) Control
Formulation protein with no preservative
Gene ID (Entrez) 1152, 1158
Name Creatine Kinase-MB Kit, Calibrator Grade
Quantity 100 μg
Source E. Coli
Regulatory Status RUO
Gene Alias Bck; B-CK; Brain creatine kinase; CK MB; Ck3; Ck-3; CKB; Ckbb; Ckbr; Ckm; CKMM; CPK-B; CPK-M; creatine kinase B; creatine kinase B chain; creatine kinase brain; creatine kinase brain-type; Creatine kinase B-type; creatine kinase M chain; Creatine kinase M-type; creatine kinase, brain; creatine kinase, M-type; creatine kinase, muscle; creatine kinase-B; creatine kinase-M; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29; HEL-211; HEL-S-29; MCK; M-CK
Common Name Creatine Kinase MB
Biological Activity Specific activity > 950 U/mg by Genzyme Manual Spectrophotometer
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Expression System E. coli
Protein Subtype Recombinant
Purity or Quality Grade 95% by SDS PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.