Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Creatine Kinase MB Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA513830
Description
This antibody is predicted to react with bovine, porcine and rabbit based on sequence homology.
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
Specifications
| Creatine Kinase MB | |
| Polyclonal | |
| Unconjugated | |
| Ckb | |
| Bck; B-CK; Brain creatine kinase; CK MB; Ck3; Ck-3; CKB; Ckbb; Ckbr; Ckm; CKMM; CPK-B; CPK-M; creatine kinase B; creatine kinase B chain; creatine kinase brain; creatine kinase brain-type; Creatine kinase B-type; creatine kinase M chain; Creatine kinase M-type; creatine kinase, brain; creatine kinase, M-type; creatine kinase, muscle; creatine kinase-B; creatine kinase-M; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29; HEL-211; HEL-S-29; MCK; M-CK | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 1152, 1158 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 1.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P06732, P12277 | |
| Ckb, CKM | |
| KLH conjugated synthetic peptide between 283-313 amino acids from the C-terminal region of human CKM. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction