Learn More
Description
Creatine (Monohydrate) is a white, odorless, crystalline powder that is sparingly soluble in water.
Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.
Alternative name(s): Creatine hydrate, Creatine monohydrate
Specifications
Specifications
| Chemical Name or Material | Creatine (Monohydrate) (Dietary Supplement Grade) |
| Packaging | Jar, 2.8L, White, HDPE, Round, 110-400, TE Pano Neck |
| CAS | 6020-87-7 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 149.1489 |
| Quantity | 1 kg |
| Percent Purity | 99 to 102 % |
| Grade | Ungraded |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
