Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatinine Amidohydrolase from Pseudomonas sp., MP Biomedical

Catalog No. MP215349183 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kU
100 U
25 U
250 U
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215349183 250 U
MP215349125 25 U
MP215349180 100 U
MP215349191 1 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP215349183 Supplier MP Biomedicals, Inc Supplier No. 0215349183
Add to Cart
Add to Cart

Creatinine Amidohydrolase from Pseudomonas sp. is useful for enzymatic determination of creatinine when coupled with creatine amidinohydrolase, sarcosine dehydrogenase or sarcosine oxidase and formaldehyde dehydrogenase in clinical analysis.

  • Creatininase from Pseudomonas sp. is a homohexameric enzyme with a molecular mass of 28.4 kDa per subunit
  • It is a cyclic amidohydrolase catalysing the reversible conversion of creatinine to creatine
  • Each monomer contains a binuclear zinc centre near the C termini of the β-strands and the N termini of the main α-helices
  • These zinc ions indicate the location of the active site
  • One unit causes the formation of one μmole of creatinine-picrate per minute at pH 7.5 at 37°C
    150 units/mg solid
TRUSTED_SUSTAINABILITY

Specifications

Color White
pH 6.5 to 7.5 (Optimum); pH Stability: pH 7.5 to 9.0 (5°C, 16hr).
Packaging Glass
Quantity 250 U
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.