Learn More
Bioassay Systems QuantiChrom™ Creatinine Assay Kit. For quantitative determination of creatinine and evaluation of drug effects on creatinine metabolism.
SDP
Catalog No. 50489213 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of creatinine and evaluation of drug effects on creatinine metabolism. Key Features: Sensitive and accurate. Use 30 µL samples. Detection limit 0.10 mg/dL (8 µM) creatinine in 96-well plate assay. Simple and high-throughput. The procedure involves addition of a single working reagent and incubation for 5 min. Can be automated as a high-throughput assay for thousands of samples per day. Improved reagent stability and versatility. The optimized formulation has greatly enhanced reagent and signal stability. Assays can be executed in 96-well plate or cuvet. Low interference in biological samples. No pretreatments are needed. Assays can be directly performed on raw biological samples. Method: OD510nm. Samples: Urine, serum, plasma and biological preparations. Species: All. Procedure: Assay takes 20 min. Kit size: 500 tests. Detection limit: 0.1 mg/dL (8 µM).
Catalog No. 50-489-213 Supplier Bioassay Systems Supplier No. DICT500
For quantitative determination of creatinine and evaluation of drug effects on creatinine metabolism. Key Features: Sensitive and accurate. Use 30 µL samples. Detection limit 0.10 mg/dL (8 µM) creatinine in 96-well plate assay. Simple and high-throughput. The procedure involves addition of a single working reagent and incubation for 5 min. Can be automated as a high-throughput assay for thousands of samples per day. Improved reagent stability and versatility. The optimized formulation has greatly enhanced reagent and signal stability. Assays can be executed in 96-well plate or cuvet. Low interference in biological samples. No pretreatments are needed. Assays can be directly performed on raw biological samples. Method: OD510nm. Samples: Urine, serum, plasma and biological preparations. Species: All. Procedure: Assay takes 20 min. Kit size: 500 tests. Detection limit: 0.1 mg/dL (8 µM).