Learn More
Ethos Biosciences Creatinine Chemical Assay Kit, 2x96 well plates, 176 samples/88 in duplicate, easy to use chemical assay - from prep to data in less than 1 hour
Supplier:  Ethos Biosciences 1012
Encompass
This assay is designed for companion use with Exocell’s immunospecific ELISAs, allowing expression of results as μg protein (i.e. albumin) per mg creatinine in the urine. This is typically used as a normalized value to compare samples. The Creatinine Companion kit contains all of the components and reagents you need to assay creatinine of any species in up to 88 duplicate urine samples. This is a chemical assay, does not contain antibodies, and is recommended only for urine.Kit Contents:. 96 well microtiter plates (2), Picrate Reagent (2x10ml), 1 N NaOH, Acid Reagent (2x12 ml), Standards (1, 3 and 10 mg/dL), & instructions.
Catalog No. NC9129340