Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatinine, NF, 98.5-102%, Spectrum™ Chemical
SDP

Catalog No. 18601867 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-867 25 g Poly Bottle
18-601-868 100 g Poly Bottle
18-601-869 500 g Poly Bottle
18-601-870 1 kg Poly Pail
18-601-871 12 kg Poly Pail
18-601-872 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-601-867 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C140325GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

C1403, 60-27-5, C4H7N3O

Spectrum™ Chemical Creatinine, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 60-27-5
Molecular Formula C4H7N3O
Molecular Weight (g/mol) 113.12
InChI Key DDRJAANPRJIHGJ-UHFFFAOYSA-N
IUPAC Name 2-amino-1-methyl-4,5-dihydro-1H-imidazol-4-one
SMILES CN1CC(=O)N=C1N

Specifications

CAS Max % 100%
Quantity 25 g
Percent Purity 98.5 to 102%
Grade NF
Ignition Residue 0.2%
Loss on Drying 3%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.