Learn More
ECM Bioscience CREB Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience CK6460
Encompass_Preferred
The CREB phospho-regulation antibody sampler kit can be used to examine Ser-133 and Ser-271 phosphorylation in CREB. The kit also includes a rabbit polyclonal antibody to monitor total expression levels of CREB, and HRP-conjugated secondary reagents. Applications: WB, E; Species Reactivity: Hu, Rt, Ms, Ck
Catalog No. 50-206-8630
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.