Learn More
Description
PA1-850 detects both the phosphorylated and non-phosphorylated forms of cyclic-AMP response element binding protein (CREB) from rat, mouse, and human samples. PA1-850 has been successfully used in ChIP, Western blot, ICC/IF and IHC-P procedures. By Western blot, this antibody detects an ~43 kDa protein representing CREB from GH4 cell extract. The PA1-850 immunogen is a synthetic peptide corresponding to residues K(123) R R E I L S R R P S Y R K(136) of human CREB. This sequence is completely conserved between human, bovine, canine, mouse, pig, sheep, and rat CREB protein. The PA1-850 immunizing peptide (Cat. # PEP-142) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | CREB |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | CREB1 |
| Gene Accession No. | P15337, P16220, Q01147 |
| Gene Alias | 2310001E10Rik; 3526402H21Rik; active transcription factor CREB; AV083133; cAMP response element binding protein 1; cAMP responsive element binding protein 1; cAMP-response element-binding protein-1; cAMP-responsive element-binding protein 1; CREB; CREB1; Creb-1; cyclic adenosine 3',5'-monophosphate response element-binding protein CREB; cyclic AMP-responsive element-binding protein 1; htCREB; MGC9284; OTTHUMP00000206661; OTTHUMP00000206662; OTTHUMP00000206667; transactivator protein; Y protein |
| Gene Symbols | CREB1, CrebB |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
