Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CREB3L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537966
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537966 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537966 Supplier Invitrogen™ Supplier No. PA537966
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with human, dog and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

CREB3L2 is a member of the old astrocyte specifically induced substance (OASIS) DNA binding and basic leucine zipper dimerization (bZIP) family of transcription factors, which includes CREB3 (MIM 606443) and CREB4 (MIM 607138).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREB3L2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Creb3l2
Gene Accession No. Q6QDP7, Q70SY1, Q8BH52
Gene Alias basic transcription factor 2; BBF2 human homolog on chromosome 7; BBF2H7; B-ZIB transcription factor; C530025K05Rik; cAMP responsive element binding protein 3 like 2; cAMP responsive element binding protein 3-like 2; cAMP-responsive element-binding protein 3-like protein 2; Creb3l2; Cyclic AMP-responsive element-binding protein 3-like protein 2; FUS/BBF2H7 protein; Processed cyclic AMP-responsive element-binding protein 3-like protein 2; Ra69; SCI-related protein Ra69; SCIRR69; TCAG_1951439
Gene Symbols Creb3l2
Host Species Goat
Immunogen Peptide with sequence C-HSLQEPYTASVVRS, from the internal region of CREB3L2 (aa 413-426).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 208647, 362339, 64764
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.